Order Entry
Ireland
ContactUsLinkComponent
Mouse TPBG, His Tag (TPG-M52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 32 - Ser 361 (Accession # Q9Z0L0-1). 1 * 1 mg
Catalog # ACRBTPG-M52H3-1MG
Supplier:  ACROBIOSYSTEMS
Mouse TPBG, His Tag (TPG-M52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 32 - Ser 361 (Accession # Q9Z0L0-1). 1 * 1 mg
Catalog # ACRBTPG-M52H3-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together