Order Entry
Ireland
ContactUsLinkComponent
Cynomolgus LILRB2, His Tag (LI2-C52H3) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Leu 448 (Accession # BAE8 7348.1). 1 * 1 mg
Catalog # ACRBLI2-C52H3-1MG
Supplier:  ACRO Biosystems
Cynomolgus LILRB2, His Tag (LI2-C52H3) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Leu 448 (Accession # BAE8 7348.1). 1 * 1 mg
Catalog # ACRBLI2-C52H3-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together