Order Entry
Ireland
ContactUsLinkComponent
Biotinylated Human TIGIT, Mouse IgG2a Fc,Avitag (TIT-H82F3) is expressed from human 293 cells (HEK293). It contains AA Met 22 - Pro 141 (Accession # Q495A1-1). 1 * 25 µG
Catalog # ACRBTIT-H82F3-25UG
Supplier:  ACRO Biosystems
Biotinylated Human TIGIT, Mouse IgG2a Fc,Avitag (TIT-H82F3) is expressed from human 293 cells (HEK293). It contains AA Met 22 - Pro 141 (Accession # Q495A1-1). 1 * 25 µG
Catalog # ACRBTIT-H82F3-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together