Order Entry
Ireland
ContactUsLinkComponent
Human CD7, Fc Tag (CD7-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1). 1 * 1 mg
Catalog # ACRBCD7-H5253-1MG
Supplier:  ACROBIOSYSTEMS
Human CD7, Fc Tag (CD7-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1). 1 * 1 mg
Catalog # ACRBCD7-H5253-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together