Order Entry
Ireland
ContactUsLinkComponent
Human EpCAM, Llama IgG2b Fc Tag (EPM-H5259) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession # AAH14785.1). 1 * 100 µG
  ACRBEPM-H5259100UG
Human EpCAM, Llama IgG2b Fc Tag (EPM-H5259) is expressed from human 293 cells (HEK293). It contains AA Gln 24 - Lys 265 (Accession # AAH14785.1). 1 * 100 µG
  ACRBEPM-H5259100UG
Frequently Bought Together