Order Entry
Ireland
ContactUsLinkComponent
Human CD5, Fc Tag (CD5-H5253) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Pro 372 (Accession # P06127-1). 1 * 1 mg
Catalog # ACRBCD5-H5253-1MG
Supplier:  ACROBIOSYSTEMS
Human CD5, Fc Tag (CD5-H5253) is expressed from human 293 cells (HEK293). It contains AA Arg 25 - Pro 372 (Accession # P06127-1). 1 * 1 mg
Catalog # ACRBCD5-H5253-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together