Order Entry
Ireland
ContactUsLinkComponent
Human FGF-9, His Tag (FG9-H52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 4 - Ser 208 (Accession # P31371-1). 1 * 100 µG
Catalog # ACRBFG9-H52H3100UG
Supplier:  ACROBIOSYSTEMS
Human FGF-9, His Tag (FG9-H52H3) is expressed from human 293 cells (HEK293). It contains AA Leu 4 - Ser 208 (Accession # P31371-1). 1 * 100 µG
Catalog # ACRBFG9-H52H3100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together