Order Entry
Ireland
ContactUsLinkComponent
Human KRAS (2-185,G12C), His Tag (KRS-H51H3) is expressed from E.coli cells. It contains AA Thr 2 - Cys 185 (Accession # P01116-2 (G 12C)). 1 * 1 mg
Catalog # ACRBKRS-H51H3-1MG
Supplier:  ACROBIOSYSTEMS
Human KRAS (2-185,G12C), His Tag (KRS-H51H3) is expressed from E.coli cells. It contains AA Thr 2 - Cys 185 (Accession # P01116-2 (G 12C)). 1 * 1 mg
Catalog # ACRBKRS-H51H3-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together