Order Entry
Ireland
ContactUsLinkComponent
Biotinylated Human B7-H7, His,Avitag™ (B77-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Asn 344 (Acces sion # Q9UM44-1). 1 * 200 µG
Catalog # ACRBB77-H82E3200UG
Supplier:  ACRO Biosystems
Biotinylated Human B7-H7, His,Avitag™ (B77-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ile 23 - Asn 344 (Acces sion # Q9UM44-1). 1 * 200 µG
Catalog # ACRBB77-H82E3200UG
Supplier:  ACRO Biosystems
Frequently Bought Together