Order Entry
Ireland
ContactUsLinkComponent
Human CD72, His Tag (CD2-H52H3) is expressed from human 293 cells (HEK293). It contains AA Arg 117 - Asp 359 (Accession # P21854-1). 1 * 1 mg
  ACRBCD2-H52H3-1MG
Human CD72, His Tag (CD2-H52H3) is expressed from human 293 cells (HEK293). It contains AA Arg 117 - Asp 359 (Accession # P21854-1). 1 * 1 mg
  ACRBCD2-H52H3-1MG
Frequently Bought Together