Order Entry
Ireland
ContactUsLinkComponent
Mouse ENPP3, His Tag (EN3-M52H3) is expressed from human 293 cells (HEK293). It contains AA Arg 46 - Ile 874 (Accession # Q6DYE8-1). 1 * 1 mg
Catalog # ACRBEN3-M52H3-1MG
Supplier:  ACROBIOSYSTEMS
Mouse ENPP3, His Tag (EN3-M52H3) is expressed from human 293 cells (HEK293). It contains AA Arg 46 - Ile 874 (Accession # Q6DYE8-1). 1 * 1 mg
Catalog # ACRBEN3-M52H3-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together