Order Entry
Ireland
ContactUsLinkComponent
Rabbit BCMA, His Tag (BCA-R52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 1 - Asn 52 (Accession # G1SFN4-1). 1 * 100 µG
Catalog # ACRBBCA-R52H4100UG
Supplier:  ACROBIOSYSTEMS
Rabbit BCMA, His Tag (BCA-R52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 1 - Asn 52 (Accession # G1SFN4-1). 1 * 100 µG
Catalog # ACRBBCA-R52H4100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together