Order Entry
Ireland
ContactUsLinkComponent
Human IL-2 R gamma, His Tag (ILA-H52H5) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Asn 254 (Accession # P31 785-1). 1 * 1 mg
Catalog # ACRBILA-H52H5-1MG
Supplier:  ACRO Biosystems
Human IL-2 R gamma, His Tag (ILA-H52H5) is expressed from human 293 cells (HEK293). It contains AA Leu 23 - Asn 254 (Accession # P31 785-1). 1 * 1 mg
Catalog # ACRBILA-H52H5-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together