Order Entry
Ireland
ContactUsLinkComponent
Mouse IL-5, His Tag (IL5-M52H3) is expressed from human 293 cells (HEK293). It contains AA Met 21 - Gly 133 (Accession # P04401-1). 1 * 1 mg
Catalog # ACRBIL5-M52H3-1MG
Supplier:  ACROBIOSYSTEMS
Mouse IL-5, His Tag (IL5-M52H3) is expressed from human 293 cells (HEK293). It contains AA Met 21 - Gly 133 (Accession # P04401-1). 1 * 1 mg
Catalog # ACRBIL5-M52H3-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together