Order Entry
Ireland
ContactUsLinkComponent
Human IL-1 beta, His Tag (ILB-H51H3) is expressed from E.coli cells. It contains AA Ala 117 - Ser 269 (Accession # P01584-1). 1 * 20 µG
Catalog # ACRBILB-H51H3-20UG
Supplier:  ACROBIOSYSTEMS
Human IL-1 beta, His Tag (ILB-H51H3) is expressed from E.coli cells. It contains AA Ala 117 - Ser 269 (Accession # P01584-1). 1 * 20 µG
Catalog # ACRBILB-H51H3-20UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together