Order Entry
Ireland
ContactUsLinkComponent
Human TSPAN8, His Tag (TS8-H52H3) is expressed from human 293 cells (HEK293). It contains AA Lys 110 - Asn 205 (Accession # P19075-1 ). 1 * 1 mg
Catalog # ACRBTS8-H52H3-1MG
Supplier:  ACROBIOSYSTEMS
Human TSPAN8, His Tag (TS8-H52H3) is expressed from human 293 cells (HEK293). It contains AA Lys 110 - Asn 205 (Accession # P19075-1 ). 1 * 1 mg
Catalog # ACRBTS8-H52H3-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together