Order Entry
Ireland
ContactUsLinkComponent
Test Sieve,350x60w0,5mm ASTM E11/FDA HAVER Test Sieve 350x60 mm - stainless steel frame AISI 304 Stainless steel metal wire cloth AISI 316 w 500 µm ASTM E11 No. 35 1 * 1 items
Catalog # HAVE208812283
Supplier:  NEXOPART
Test Sieve,350x60w0,5mm ASTM E11/FDA HAVER Test Sieve 350x60 mm - stainless steel frame AISI 304 Stainless steel metal wire cloth AISI 316 w 500 µm ASTM E11 No. 35 1 * 1 items
Catalog # HAVE208812283
Supplier:  NEXOPART
Frequently Bought Together