Order Entry
Ireland
ContactUsLinkComponent
Biotinylated SARS-CoV-2 Spike RBD (L452R, T478K), His,Avitag (SPD-C82Ed) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession QHD43416.1 (L452R, T478K)). The mutations (L452R, T478K) were identified in the SARS-CoV 1 * 200 µG
Catalog # ACRBSPD-C82ED200UG
Supplier:  ACRO Biosystems
Biotinylated SARS-CoV-2 Spike RBD (L452R, T478K), His,Avitag (SPD-C82Ed) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession QHD43416.1 (L452R, T478K)). The mutations (L452R, T478K) were identified in the SARS-CoV 1 * 200 µG
Catalog # ACRBSPD-C82ED200UG
Supplier:  ACRO Biosystems
Frequently Bought Together