Order Entry
Ireland
ContactUsLinkComponent
Vial & Cap Kit. 100 EA 2ml, Screw Top, Hydrophobic, Clear Autosampler Vials and matching Black Caps with Clear Silicone Rubber / PTF E Septa. RSA-Pro X Brand. 1 PK. 1 * 100 items
Catalog # MICS9509S-1DCP-PX
Supplier:  MICROSOLV
Vial & Cap Kit. 100 EA 2ml, Screw Top, Hydrophobic, Clear Autosampler Vials and matching Black Caps with Clear Silicone Rubber / PTF E Septa. RSA-Pro X Brand. 1 PK. 1 * 100 items
Catalog # MICS9509S-1DCP-PX
Supplier:  MICROSOLV
Frequently Bought Together