Order Entry
Ireland
ContactUsLinkComponent
Gas-tight suit Tychem TK with Socks. For use with SCBA. Attached socks. Attached two-layer glove system. Wide panoramic visor. Gas-tight zipper fitted to the front side of the suit. Yellow. Size L 1 * 1 items
Catalog # DUPOTKGEVHTYL00/L
Supplier:  DuPont
Gas-tight suit Tychem TK with Socks. For use with SCBA. Attached socks. Attached two-layer glove system. Wide panoramic visor. Gas-tight zipper fitted to the front side of the suit. Yellow. Size L 1 * 1 items
Catalog # DUPOTKGEVHTYL00/L
Supplier:  DuPont