Order Entry
Ireland
ContactUsLinkComponent
AREG ELISA Kit (Human)is based on standard sandwich enzyme-linked immuno-sorbent assay technology. An antibody specific for AREG has been pre-coated onto a 96-well plate (12 x 8 Well Strips) and blocked. Standards or test samples are added to the wel 1 * 1 KIT
Catalog # AVIVOKCD05603
AREG ELISA Kit (Human)is based on standard sandwich enzyme-linked immuno-sorbent assay technology. An antibody specific for AREG has been pre-coated onto a 96-well plate (12 x 8 Well Strips) and blocked. Standards or test samples are added to the wel 1 * 1 KIT
Catalog # AVIVOKCD05603
Frequently Bought Together