Order Entry
Ireland
ContactUsLinkComponent
Human CD32b, His Tag SPR  BLI verified CDB-H5228 is expressed from human 293 cells HEK293. It contains AA Ala 46 - Pro 217 Accession P31994. 1 * 50 µG
Catalog # ACRBCDB-H5228-50UG
Supplier:  ACRO Biosystems
Human CD32b, His Tag SPR  BLI verified CDB-H5228 is expressed from human 293 cells HEK293. It contains AA Ala 46 - Pro 217 Accession P31994. 1 * 50 µG
Catalog # ACRBCDB-H5228-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together