Order Entry
Ireland
ContactUsLinkComponent
Rhesus macaque Properdin, His Tag PRN-R52H6 is expressed from human 293 cells HEK293. It contains AA Asp 28 - Leu 469 Accession F7C GG5-1. 1 * 100 µG
Catalog # ACRBPRN-R52H6100UG
Supplier:  ACRO Biosystems
Rhesus macaque Properdin, His Tag PRN-R52H6 is expressed from human 293 cells HEK293. It contains AA Asp 28 - Leu 469 Accession F7C GG5-1. 1 * 100 µG
Catalog # ACRBPRN-R52H6100UG
Supplier:  ACRO Biosystems
Frequently Bought Together