Order Entry
Ireland
ContactUsLinkComponent
Cynomolgus TSLP, His Tag TSP-C52H8 is expressed from human 293 cells HEK293. It contains AA Tyr 29 - Gln 159 Accession A0A2K5TXV0-1 . 1 * 1 mg
Catalog # ACRBTSP-C52H8-1MG
Supplier:  ACROBIOSYSTEMS
Cynomolgus TSLP, His Tag TSP-C52H8 is expressed from human 293 cells HEK293. It contains AA Tyr 29 - Gln 159 Accession A0A2K5TXV0-1 . 1 * 1 mg
Catalog # ACRBTSP-C52H8-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together