Order Entry
Ireland
ContactUsLinkComponent
Biotinylated Human Cynomolgus Rhesus macaque CD28, His,Avitag CD8-H82E5 is expressed from human 293 cells HEK293. It contains AA A sn 19 - Pro 152 Accession P10747-1. 1 * 200 µG
Catalog # ACRBCD8-H82E5200UG
Supplier:  ACRO Biosystems
Biotinylated Human Cynomolgus Rhesus macaque CD28, His,Avitag CD8-H82E5 is expressed from human 293 cells HEK293. It contains AA A sn 19 - Pro 152 Accession P10747-1. 1 * 200 µG
Catalog # ACRBCD8-H82E5200UG
Supplier:  ACRO Biosystems
Frequently Bought Together