Order Entry
Ireland
ContactUsLinkComponent
Anti-SARS-CoV-2 Nucleocapsid Antibody, Human IgG1 NUN-S41 is isolated from a SARS-CoV-2 infected patient and is recombinantly produc ed from human 293 cells HEK293. 1 * 1 mg
Catalog # ACRBNUN-S41-1MG
Supplier:  ACROBIOSYSTEMS
Anti-SARS-CoV-2 Nucleocapsid Antibody, Human IgG1 NUN-S41 is isolated from a SARS-CoV-2 infected patient and is recombinantly produc ed from human 293 cells HEK293. 1 * 1 mg
Catalog # ACRBNUN-S41-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together