Order Entry
Ireland
ContactUsLinkComponent
SARS-CoV-2 S protein RBD, His Tag SPD-C52H3 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Lys 537 Accession QH D43416.1. 1 * 1 mg
Catalog # ACRBSPD-C52H3-1MG
Supplier:  ACRO Biosystems
SARS-CoV-2 S protein RBD, His Tag SPD-C52H3 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Lys 537 Accession QH D43416.1. 1 * 1 mg
Catalog # ACRBSPD-C52H3-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together