Order Entry
Ireland
ContactUsLinkComponent
SARS-CoV-2 S2 protein, His Tag S2N-C52H5 is expressed from human 293 cells HEK293. It contains AA Ser 686 - Pro 1213 Accession QHD4 3416.1. Proline substitutions F817P, A892P, A899P, A942P, K986P, V987P are introduced to prevent the formation of aggr 1 * 1 mg
Catalog # ACRBS2N-C52H5-1MG
Supplier:  ACROBIOSYSTEMS
SARS-CoV-2 S2 protein, His Tag S2N-C52H5 is expressed from human 293 cells HEK293. It contains AA Ser 686 - Pro 1213 Accession QHD4 3416.1. Proline substitutions F817P, A892P, A899P, A942P, K986P, V987P are introduced to prevent the formation of aggr 1 * 1 mg
Catalog # ACRBS2N-C52H5-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together