Order Entry
Ireland
ContactUsLinkComponent
Mouse GUCY2C, His Tag GUC-M52H3 is expressed from human 293 cells HEK293. It contains AA Val 20 - Met 433 Accession Q3UWA6-1. 1 * 1 mg
Catalog # ACRBGUC-M52H3-1MG
Supplier:  ACROBIOSYSTEMS
Mouse GUCY2C, His Tag GUC-M52H3 is expressed from human 293 cells HEK293. It contains AA Val 20 - Met 433 Accession Q3UWA6-1. 1 * 1 mg
Catalog # ACRBGUC-M52H3-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together