Order Entry
Ireland
ContactUsLinkComponent
Cynomolgus CD20 Full Length, His Tag CD0-C52H8 is expressed from human 293 cells HEK293. It contains AA Met 1 - Pro 297 Accession M 4ZHZ6-1 . 1 * 500 µG
Catalog # ACRBCD0-C52H8500UG
Supplier:  ACRO Biosystems
Cynomolgus CD20 Full Length, His Tag CD0-C52H8 is expressed from human 293 cells HEK293. It contains AA Met 1 - Pro 297 Accession M 4ZHZ6-1 . 1 * 500 µG
Catalog # ACRBCD0-C52H8500UG
Supplier:  ACRO Biosystems
Frequently Bought Together