Order Entry
Ireland
ContactUsLinkComponent
Biotinylated Onchocerca volvulus OV16 protein, Avi,GST Tag OV6-V81G9 is expressed from E.coli cells. It contains AA Lys 17 - Asp 197 Accession P31729-1. 1 * 200 µG
Catalog # ACRBOV6-V81G9200UG
Supplier:  ACRO Biosystems
Biotinylated Onchocerca volvulus OV16 protein, Avi,GST Tag OV6-V81G9 is expressed from E.coli cells. It contains AA Lys 17 - Asp 197 Accession P31729-1. 1 * 200 µG
Catalog # ACRBOV6-V81G9200UG
Supplier:  ACRO Biosystems
Frequently Bought Together