Order Entry
Ireland
ContactUsLinkComponent
Human RNF43, Fc Tag RN3-H5256 is expressed from human 293 cells HEK293. It contains AA Gln 44 - Tyr 197 Accession Q68DV7-1. 1 * 1 mg
Catalog # ACRBRN3-H5256-1MG
Supplier:  ACROBIOSYSTEMS
Human RNF43, Fc Tag RN3-H5256 is expressed from human 293 cells HEK293. It contains AA Gln 44 - Tyr 197 Accession Q68DV7-1. 1 * 1 mg
Catalog # ACRBRN3-H5256-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together