Order Entry
Ireland
ContactUsLinkComponent
FITC-Labeled Human CD38, His Tag Cat. No. CD8-HF2H5 is expressed from human HEK293 cells. It contains AA Val 43 - Ile 300 Accession NP_001766. It is the FITC labeled form of Human CD38 Protein, His Tag Cat. No. CD8-H5224. 1 * 25 µG
Catalog # ACRBCD8-HF2H5-25UG
Supplier:  ACRO Biosystems
FITC-Labeled Human CD38, His Tag Cat. No. CD8-HF2H5 is expressed from human HEK293 cells. It contains AA Val 43 - Ile 300 Accession NP_001766. It is the FITC labeled form of Human CD38 Protein, His Tag Cat. No. CD8-H5224. 1 * 25 µG
Catalog # ACRBCD8-HF2H5-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together