Biotinylated SARS-CoV-2 S protein D614G, His,Avitag, Super stable trimer SPN-C82E3 is the ectodomain of SARS-CoV-2 S protein that co ntains AA Val 16 - Pro 1213 Accession QHD43416.1 D614G and D614G mutation, which has become increasingly common in S 1 * 200 µG
Catalog # ACRBSPN-C82E3200UG
Supplier: ACROBIOSYSTEMS
Frequently Bought Together