Order Entry
Ireland
ContactUsLinkComponent
FITC-Labeled Human EpCAM, His Tag EPM-HF225 is expressed from human 293 cells HEK293. It contains AA Gln 24 - Lys 265 Accession AAH 14785.1. 1 * 25 µG
Catalog # ACRBEPM-HF225-25UG
Supplier:  ACROBIOSYSTEMS
FITC-Labeled Human EpCAM, His Tag EPM-HF225 is expressed from human 293 cells HEK293. It contains AA Gln 24 - Lys 265 Accession AAH 14785.1. 1 * 25 µG
Catalog # ACRBEPM-HF225-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together