Order Entry
Ireland
ContactUsLinkComponent
Human PLGF 19-149, His Tag PGF-H52H5 is expressed from human 293 cells HEK293. It contains AA Leu 19 - Arg 149 Accession P49763-2. 1 * 500 µG
Catalog # ACRBPGF-H52H5500UG
Supplier:  ACRO Biosystems
Human PLGF 19-149, His Tag PGF-H52H5 is expressed from human 293 cells HEK293. It contains AA Leu 19 - Arg 149 Accession P49763-2. 1 * 500 µG
Catalog # ACRBPGF-H52H5500UG
Supplier:  ACRO Biosystems
Frequently Bought Together