Order Entry
Ireland
ContactUsLinkComponent
Mouse TGFB1 Protein TG1-M5218 is expressed from human 293 cells HEK293. It contains AA Ala 279 - Ser 390 Accession P04202-1. 1 * 50 µG
Catalog # ACRBTG1-M5218-50UG
Supplier:  ACROBIOSYSTEMS
Mouse TGFB1 Protein TG1-M5218 is expressed from human 293 cells HEK293. It contains AA Ala 279 - Ser 390 Accession P04202-1. 1 * 50 µG
Catalog # ACRBTG1-M5218-50UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together