Order Entry
Ireland
ContactUsLinkComponent
Biotinylated Human IL-3 R alpha, His,Avitag ILA-H82H5 is expressed from human 293 cells HEK293. It contains AA Thr 19 - Arg 305 Acce ssion P26951-1. 1 * 25 µG
Catalog # ACRBILA-H82H5-25UG
Supplier:  ACRO Biosystems
Biotinylated Human IL-3 R alpha, His,Avitag ILA-H82H5 is expressed from human 293 cells HEK293. It contains AA Thr 19 - Arg 305 Acce ssion P26951-1. 1 * 25 µG
Catalog # ACRBILA-H82H5-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together