Order Entry
Ireland
ContactUsLinkComponent
Biotinylated Mouse IgG1 Fc, Avitag™ IG1-M8211 is expressed from human 293 cells HEK293. It contains AA Val 98 - Lys 324 Accession A AK53870.1. 1 * 200 µG
Catalog # ACRBIG1-M8211200UG
Supplier:  ACRO Biosystems
Biotinylated Mouse IgG1 Fc, Avitag™ IG1-M8211 is expressed from human 293 cells HEK293. It contains AA Val 98 - Lys 324 Accession A AK53870.1. 1 * 200 µG
Catalog # ACRBIG1-M8211200UG
Supplier:  ACRO Biosystems
Frequently Bought Together