Order Entry
Ireland
ContactUsLinkComponent
SARS-CoV-2 S protein RBD E484K, His Tag SRD-C52H3 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Lys 537 Accessi on QHD43416.1 E484K. E484K mutaion was identified in the SARS-CoV-2 variant known as 20C501Y.V2 or B.1.351 lineage w 1 * 1 mg
Catalog # ACRBSRD-C52H3-1MG
Supplier:  ACRO Biosystems
SARS-CoV-2 S protein RBD E484K, His Tag SRD-C52H3 is expressed from human 293 cells HEK293. It contains AA Arg 319 - Lys 537 Accessi on QHD43416.1 E484K. E484K mutaion was identified in the SARS-CoV-2 variant known as 20C501Y.V2 or B.1.351 lineage w 1 * 1 mg
Catalog # ACRBSRD-C52H3-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together