Order Entry
Ireland
ContactUsLinkComponent
Human 4-1BB, His Tag 41B-H52Hc is expressed from human 293 cells HEK293. It contains AA Leu 24 - Gln 186 Accession Q07011-1. 1 * 1 mg
Catalog # ACRB41B-H52HC-1MG
Supplier:  ACROBIOSYSTEMS
Human 4-1BB, His Tag 41B-H52Hc is expressed from human 293 cells HEK293. It contains AA Leu 24 - Gln 186 Accession Q07011-1. 1 * 1 mg
Catalog # ACRB41B-H52HC-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together