Order Entry
Ireland
ContactUsLinkComponent
Human ENPP3, His Tag EN3-H52H4 is expressed from human 293 cells HEK293. It contains AA Leu 48 - Ile 875 Accession O14638-1. 1 * 1 mg
Catalog # ACRBEN3-H52H4-1MG
Supplier:  ACROBIOSYSTEMS
Human ENPP3, His Tag EN3-H52H4 is expressed from human 293 cells HEK293. It contains AA Leu 48 - Ile 875 Accession O14638-1. 1 * 1 mg
Catalog # ACRBEN3-H52H4-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together