Order Entry
Ireland
ContactUsLinkComponent
Mouse Nectin-4, His Tag NE4-M52Ha is expressed from human 293 cells HEK293. It contains AA Gly 31 - Ser 347 Accession Q8R007-1. 1 * 100 µG
  ACRBNE4-M52HA100UG
Mouse Nectin-4, His Tag NE4-M52Ha is expressed from human 293 cells HEK293. It contains AA Gly 31 - Ser 347 Accession Q8R007-1. 1 * 100 µG
  ACRBNE4-M52HA100UG
Frequently Bought Together