Order Entry
Ireland
ContactUsLinkComponent
Human PVRIG, His Tag PVG-H52H5 is expressed from human 293 cells HEK293. It contains AA Thr 41 - Asp 171 Accession Q6DKI7-1. 1 * 1 mg
Catalog # ACRBPVG-H52H5-1MG
Supplier:  ACROBIOSYSTEMS
Human PVRIG, His Tag PVG-H52H5 is expressed from human 293 cells HEK293. It contains AA Thr 41 - Asp 171 Accession Q6DKI7-1. 1 * 1 mg
Catalog # ACRBPVG-H52H5-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together