Order Entry
Ireland
ContactUsLinkComponent
Anti-SARS-CoV-2 Spike S2 protein Antibody, Human IgG4 AS86 is isolated from a SARS-CoV-2 infected patient and is recombinantly produ ced from CHO cells. 1 * 1 mg
Catalog # ACRBS2N-S86-1MG
Supplier:  ACRO Biosystems
Anti-SARS-CoV-2 Spike S2 protein Antibody, Human IgG4 AS86 is isolated from a SARS-CoV-2 infected patient and is recombinantly produ ced from CHO cells. 1 * 1 mg
Catalog # ACRBS2N-S86-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together