Order Entry
Ireland
ContactUsLinkComponent
Human CD45, His Tag CD5-H52H8 is expressed from human 293 cells HEK293. It contains AA Gln 26 - Lys 577 Accession P08575-3. 1 * 1 mg
Catalog # ACRBCD5-H52H8-1MG
Supplier:  ACROBIOSYSTEMS
Human CD45, His Tag CD5-H52H8 is expressed from human 293 cells HEK293. It contains AA Gln 26 - Lys 577 Accession P08575-3. 1 * 1 mg
Catalog # ACRBCD5-H52H8-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together