SARS-CoV-2 S1 protein HV69-70del, His Tag S1N-C52Hd is expressed from human 293 cells HEK293. It contains AA Val 16 - Arg 685 Access ion QHD43416.1 HV69-70del. HV69-70 deletion was identified in the SARS-CoV-2 variant known as 20B501Y.V1, VOC 202012 1 * 1 mg
Catalog # ACRBS1N-C52HD-1MG
Supplier: ACRO Biosystems
Frequently Bought Together