Order Entry
Ireland
ContactUsLinkComponent
Human IL-11, His Tag IL1-H5243 is expressed from human 293 cells HEK293. It contains AA Pro 22 - Leu 199 Accession P20809-1. 1 * 100 µG
  ACRBIL1-H5243100UG
Human IL-11, His Tag IL1-H5243 is expressed from human 293 cells HEK293. It contains AA Pro 22 - Leu 199 Accession P20809-1. 1 * 100 µG
  ACRBIL1-H5243100UG
Frequently Bought Together