Order Entry
Ireland
ContactUsLinkComponent
Human HBEGF, His Tag HBF-H52H3 is expressed from human 293 cells HEK293. It contains AA Asp 63 - Leu 148 Accession Q99075-1. 1 * 1 mg
Catalog # ACRBHBF-H52H3-1MG
Supplier:  ACROBIOSYSTEMS
Human HBEGF, His Tag HBF-H52H3 is expressed from human 293 cells HEK293. It contains AA Asp 63 - Leu 148 Accession Q99075-1. 1 * 1 mg
Catalog # ACRBHBF-H52H3-1MG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together