Order Entry
Ireland
ContactUsLinkComponent
Rat CLEC12A, His Tag CLA-R5248 is expressed from human 293 cells HEK293. It contains AA Tyr 65 - Leu 269 Accession B4F798-1. 1 * 100 µG
Catalog # ACRBCLA-R5248100UG
Supplier:  ACRO Biosystems
Rat CLEC12A, His Tag CLA-R5248 is expressed from human 293 cells HEK293. It contains AA Tyr 65 - Leu 269 Accession B4F798-1. 1 * 100 µG
Catalog # ACRBCLA-R5248100UG
Supplier:  ACRO Biosystems
Frequently Bought Together